Assay Design



Gene Transcripts

Gene Symbol
rpusd4
Entrez Gene ID
445101
Gene Name
RNA pseudouridine synthase D4
Gene Aliases
si:ch211-14i3.3, zgc:92006
Chromosome Location
Chr.11: 369407 - 380240 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003495.1 NP_001003495.1 6 - 7 1018 122
GenBank mRNA BC078405.1 - 6 - 7 1018 122
BC152262.1 - 6 - 7 1066 122

Gene Symbol
LOC110438921
Entrez Gene ID
110438921
Gene Name
RNA pseudouridylate synthase domain-containing protein 4-like
Gene Aliases
-
Chromosome Location
Chr.11: 404 - 5577 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021479776.1 XP_021335451.1 2 - 3 285 122

View Full Product Details