Assay Design



Gene Transcripts

Gene Symbol
gtf2h1
Entrez Gene ID
447857
Gene Name
general transcription factor IIH, polypeptide 1
Gene Aliases
zgc:92126
Chromosome Location
Chr.25: 3196855 - 3217115 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004596.1 NP_001004596.1 13 - 14 1615 79
GenBank mRNA BC081602.1 - 13 - 14 1615 79

View Full Product Details