Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005402.1 | NP_001005402.1 | 17 - 18 | 2089 | 128 |
XM_005156820.4 | XP_005156877.1 | 16 - 17 | 2202 | 128 | |
GenBank mRNA | AY684930.1 | - | 17 - 18 | 2089 | 128 |