Assay Design



Gene Transcripts

Gene Symbol
glmna
Entrez Gene ID
492774
Gene Name
glomulin, FKBP associated protein a
Gene Aliases
glmnl, zgc:101567
Chromosome Location
Chr.2: 10821066 - 10857900 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007415.1 NP_001007416.1 2 - 3 293 94
XM_005171163.4 XP_005171220.1 2 - 3 154 94
XM_021480037.1 XP_021335712.1 2 - 3 272 94
GenBank mRNA BC085370.1 - 2 - 3 293 94
BC165850.1 - 1 - 2 72 94

View Full Product Details