Assay Design



Gene Transcripts

Gene Symbol
ptgr1
Entrez Gene ID
494108
Gene Name
prostaglandin reductase 1
Gene Aliases
ltb4dh, wu:fj35e05, zgc:101689
Chromosome Location
Chr.7: 25816 - 31972 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008651.1 NP_001008651.1 2 - 3 191 84
GenBank mRNA BC086722.1 - 2 - 3 191 84

View Full Product Details