Assay Design



Gene Transcripts

Gene Symbol
sh2d5
Entrez Gene ID
541529
Gene Name
SH2 domain containing 5
Gene Aliases
fa11f09, wu:fa11f09, zgc:113447
Chromosome Location
Chr.11: 37638968 - 37650969 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001362556.1 NP_001349485.1 8 - 9 988 59
NM_001362557.1 NP_001349486.1 8 - 9 991 59
NR_155759.1 8 - 9 1037 59
NR_155760.1 8 - 9 1040 59
GenBank mRNA BC091828.1 - 8 - 9 1027 59

View Full Product Details