Assay Design



Gene Transcripts

Gene Symbol
zfpm2b
Entrez Gene ID
568464
Gene Name
zinc finger protein, FOG family member 2b
Gene Aliases
FOG-B, fog2b
Chromosome Location
Chr.19: 13193215 - 13286738 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039524.1 NP_001034613.1 6 - 7 992 56
GenBank mRNA DQ015977.1 - 6 - 7 992 56

View Full Product Details