Assay Design



Gene Transcripts

Gene Symbol
f9b
Entrez Gene ID
678552
Gene Name
coagulation factor IXb
Gene Aliases
zgc:136807
Chromosome Location
Chr.10: 2200 - 11891 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040310.1 NP_001035400.1 3 - 4 304 118
GenBank mRNA BC115275.1 - 3 - 4 304 118

View Full Product Details