Assay Design



Gene Transcripts

Gene Symbol
zcchc24
Entrez Gene ID
562028
Gene Name
zinc finger, CCHC domain containing 24
Gene Aliases
fc45d05, wu:fc45d05, zgc:158323
Chromosome Location
Chr.13: 30579260 - 30645965 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098388.2 NP_001091858.1 2 - 3 683 72
GenBank mRNA BC141714.1 - 2 - 3 612 72
CT621901.2 - 2 - 3 233 72

View Full Product Details