Assay Design



Gene Transcripts

Gene Symbol
ftr54
Entrez Gene ID
799511
Gene Name
finTRIM family, member 54
Gene Aliases
zgc:165577
Chromosome Location
Chr.10: 25819996 - 25823194 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099428.1 NP_001092898.1 1 - 2 594 147
GenBank mRNA BC142875.1 - 1 - 2 594 147

View Full Product Details