Assay Design



Gene Transcripts

Gene Symbol
dcun1d1
Entrez Gene ID
327232
Gene Name
DCN1, defective in cullin neddylation 1, domain containing 1 (S. cerevisiae)
Gene Aliases
fd19a01, wu:fd19a01, zgc:66414
Chromosome Location
Chr.2: 7698446 - 7711498 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199772.1 NP_956066.1 4 - 5 563 137
GenBank mRNA BC057530.1 - 4 - 5 563 137
BC068381.1 - 4 - 5 529 137

View Full Product Details