Assay Design



Gene Transcripts

Gene Symbol
anp32a
Entrez Gene ID
335907
Gene Name
acidic (leucine-rich) nuclear phosphoprotein 32 family, member A
Gene Aliases
wu:fj36e08, zgc:55516
Chromosome Location
Chr.7: 33345627 - 33350002 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199986.1 NP_956280.1 4 - 5 611 86
GenBank mRNA BC048047.1 - 4 - 5 611 86
BC071357.1 - 4 - 5 666 86
BC165448.1 - 4 - 5 558 86

View Full Product Details