Assay Design



Gene Transcripts

Gene Symbol
arpc5la
Entrez Gene ID
393550
Gene Name
actin related protein 2/3 complex, subunit 5-like, a
Gene Aliases
arpc5l, zgc:65881, zgc:77408
Chromosome Location
Chr.8: 41453752 - 41462734 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200578.2 NP_956872.1 2 - 3 399 73
GenBank mRNA BC056697.1 - 2 - 3 415 73
BC065664.1 - 2 - 3 399 73

View Full Product Details