Assay Design
Gene Transcripts
- Gene Symbol
- med30
- Entrez Gene ID
-
393690
- Gene Name
- mediator complex subunit 30
- Gene Aliases
-
thrap6, zgc:73113
- Chromosome Location
-
Chr.19: 46372031 - 46411359 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200717.1 |
NP_957011.1 |
3 - 4 |
412 |
111 |
GenBank mRNA |
BC059478.1 |
- |
3 - 4 |
412 |
111 |
View Full Product Details