Assay Design
Gene Transcripts
- Gene Symbol
- znrf1
- Entrez Gene ID
-
393844
- Gene Name
- zinc and ring finger 1
- Gene Aliases
-
zgc:77896
- Chromosome Location
-
Chr.25: 13406069 - 13471288 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200870.1 |
NP_957164.1 |
3 - 4 |
1106 |
63 |
GenBank mRNA |
BC063239.1 |
- |
3 - 4 |
1106 |
63 |
View Full Product Details