Assay Design



Gene Transcripts

Gene Symbol
gnai1
Entrez Gene ID
393946
Gene Name
guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 1
Gene Aliases
zgc:63957
Chromosome Location
Chr.4: 22680487 - 22714063 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200971.1 NP_957265.1 7 - 8 1142 56
GenBank mRNA BC053164.1 - 7 - 8 1142 56
BC154335.1 - 7 - 8 1001 56

View Full Product Details