Assay Design
Gene Transcripts
- Gene Symbol
- ralbp1
- Entrez Gene ID
-
404612
- Gene Name
- ralA binding protein 1
- Gene Aliases
-
zgc:76867
- Chromosome Location
-
Chr.2: 52809981 - 52841561 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_207080.1 |
NP_996963.1 |
2 - 3 |
360 |
73 |
GenBank mRNA |
BC066523.1 |
- |
2 - 3 |
360 |
73 |
View Full Product Details