Assay Design



Gene Transcripts

Gene Symbol
heg1
Entrez Gene ID
406202
Gene Name
heart development protein with EGF-like domains 1
Gene Aliases
heg
Chromosome Location
Chr.9: 38684871 - 38707510 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212968.2 NP_998133.2 1 - 2 421 75
GenBank mRNA AY507658.1 - 1 - 2 422 75
AY507659.1 - 1 - 2 422 75
AY507660.1 - 1 - 2 422 75
BC162109.1 - 1 - 2 420 75
BC162480.1 - 1 - 2 420 75

View Full Product Details