Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_213039.1 | NP_998204.1 | 3 - 4 | 386 | 59 |
XM_005171826.4 | XP_005171883.1 | 3 - 4 | 410 | 59 | |
XM_021476056.1 | XP_021331731.1 | 3 - 4 | 574 | 59 | |
XM_021476057.1 | XP_021331732.1 | 3 - 4 | 568 | 59 | |
GenBank mRNA | BC059673.1 | - | 3 - 4 | 386 | 59 |