Assay Design
Gene Transcripts
- Gene Symbol
- ccng2
- Entrez Gene ID
-
266794
- Gene Name
- cyclin G2
- Gene Aliases
-
cb438, wu:fa08f07, wu:fk28c06, wu:fk91a03, zgc:77125
- Chromosome Location
-
Chr.21: 5800354 - 5807559 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_213172.1 |
NP_998337.1 |
4 - 5 |
555 |
66 |
GenBank mRNA |
BC066368.1 |
- |
4 - 5 |
555 |
66 |
View Full Product Details