Assay Design
Gene Transcripts
- Gene Symbol
- cbwd
- Entrez Gene ID
-
406536
- Gene Name
- COBW domain containing
- Gene Aliases
-
fb76b04, fc73a04, wu:fb76b04, wu:fc73a04, zgc:77617
- Chromosome Location
-
Chr.8: 29378 - 53722 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_213253.1 |
NP_998418.1 |
15 - 16 |
1154 |
89 |
GenBank mRNA |
BC065429.1 |
- |
15 - 16 |
1154 |
89 |
View Full Product Details