Assay Design



Gene Transcripts

Gene Symbol
cab39l1
Entrez Gene ID
406822
Gene Name
calcium binding protein 39, like 1
Gene Aliases
cab39, wu:fb72b01, wu:fi37c08, zgc:55451
Chromosome Location
Chr.14: 35103 - 45476 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213501.1 NP_998666.1 6 - 7 879 68
GenBank mRNA BC044172.1 - 6 - 7 879 68
BC164384.1 - 5 - 6 660 68

View Full Product Details