Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080689.1 | NP_001074158.1 | 11 - 11 | 2051 | 87 |
NM_001347739.1 | NP_001334668.1 | 13 - 13 | 2557 | 87 | |
XM_009305034.3 | XP_009303309.2 | 13 - 13 | 2605 | 87 | |
GenBank mRNA | BC129212.1 | - | 11 - 11 | 2051 | 87 |