Assay Design
Gene Transcripts
- Gene Symbol
- setd7
- Entrez Gene ID
-
436729
- Gene Name
- SET domain containing 7, histone lysine methyltransferase
- Gene Aliases
-
set7, zgc:101860, zgc:92330
- Chromosome Location
-
Chr.14: 46199941 - 46213584 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC081418.1 |
- |
2 - 2 |
945 |
134 |
View Full Product Details