Assay Design



Gene Transcripts

Gene Symbol
wnt4
Entrez Gene ID
30123
Gene Name
wingless-type MMTV integration site family, member 4
Gene Aliases
wnt-4, wnt4a, zgc:136737
Chromosome Location
Chr.11: 39167846 - 39202915 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040387.1 NP_001035477.1 5 - 5 2054 119
GenBank mRNA BC115247.1 - 5 - 5 2054 119

View Full Product Details