Assay Design



Gene Transcripts

Gene Symbol
lrrc15
Entrez Gene ID
791200
Gene Name
leucine rich repeat containing 15
Gene Aliases
cb894, zgc:158286
Chromosome Location
Chr.6: 105231 - 108953 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080682.1 NP_001074151.1 2 - 2 1646 134
GenBank mRNA BC129189.1 - 2 - 2 1646 134

View Full Product Details