Assay Design



Gene Transcripts

Gene Symbol
fam83d
Entrez Gene ID
492689
Gene Name
family with sequence similarity 83 member D
Gene Aliases
im:7149534, zgc:162965
Chromosome Location
Chr.11: 25328202 - 25336918 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089355.1 NP_001082824.1 4 - 4 2248 129
GenBank mRNA BC139679.1 - 4 - 4 2248 129

View Full Product Details