Assay Design



Gene Transcripts

Gene Symbol
wbp4
Entrez Gene ID
553723
Gene Name
WW domain binding protein 4
Gene Aliases
zgc:112384
Chromosome Location
Chr.6: 60018233 - 60020229 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020694.1 NP_001018530.1 - 754 95
GenBank mRNA BC095798.1 - - 754 95
BC165793.1 - - 745 95

View Full Product Details