Assay Design



Gene Transcripts

Gene Symbol
pcdh2g6
Entrez Gene ID
554054
Gene Name
protocadherin 2 gamma 6
Gene Aliases
-
Chromosome Location
Chr.14: 1889679 - 2073488 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024179.1 NP_001019350.1 1 - 1 52 122
GenBank mRNA AY583054.1 - 1 - 1 52 122

View Full Product Details