Assay Design



Gene Transcripts

Gene Symbol
nog2
Entrez Gene ID
30185
Gene Name
noggin 2
Gene Aliases
zNog2
Chromosome Location
Chr.24: 38814738 - 38816725 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130992.2 NP_571067.1 1 - 1 1604 106
GenBank mRNA AF159148.1 - 1 - 1 1645 106
BC056600.1 - 1 - 1 1604 106

View Full Product Details