Assay Design
Gene Transcripts
- Gene Symbol
- pax7a
- Entrez Gene ID
-
30587
- Gene Name
- paired box 7a
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 50661 - 168175 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF014367.1 |
- |
5 - 5 |
1948 |
110 |
View Full Product Details