Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175082.3 | NP_778257.2 | 4 - 4 | 1249 | 108 |
GenBank mRNA | AY134491.1 | - | 4 - 4 | 1155 | 108 |
BC065978.1 | - | 4 - 4 | 1249 | 108 | |
BC159237.1 | - | 4 - 4 | 1115 | 108 |