Assay Design



Gene Transcripts

Gene Symbol
cldn8
Entrez Gene ID
445278
Gene Name
claudin 8
Gene Aliases
wu:fa01e05, zgc:91900
Chromosome Location
Chr.15: 42582685 - 42584552 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003733.1 NP_001003733.1 1 - 1 648 114
GenBank mRNA BC077161.1 - 1 - 1 648 114
BC164840.1 - 1 - 1 663 114

View Full Product Details