Assay Design



Gene Transcripts

Gene Symbol
thoc3
Entrez Gene ID
445302
Gene Name
THO complex 3
Gene Aliases
zgc:100815
Chromosome Location
Chr.14: 25505474 - 25510955 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003758.1 NP_001003758.1 1 - 2 196 73
GenBank mRNA BC079495.1 - 1 - 2 196 73
BC152622.1 - 1 - 2 190 73

View Full Product Details