Assay Design



Gene Transcripts

Gene Symbol
ncaldb
Entrez Gene ID
445319
Gene Name
neurocalcin delta b
Gene Aliases
ncald, zgc:100982
Chromosome Location
Chr.19: 12295443 - 12324540 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003776.1 NP_001003776.1 3 - 4 590 75
XM_005159585.3 XP_005159642.1 3 - 4 676 75
XM_005159586.4 XP_005159643.1 4 - 5 637 75
XM_009294100.3 XP_009292375.1 4 - 5 714 75
GenBank mRNA BC079490.1 - 3 - 4 590 75

View Full Product Details