Assay Design



Gene Transcripts

Gene Symbol
ing5b
Entrez Gene ID
368450
Gene Name
inhibitor of growth family, member 5b
Gene Aliases
ING1, p33, si:busm1-40g1.1, si:dz40g1.1, wu:fc94a02, zgc:100910
Chromosome Location
Chr.2: 36605994 - 36608760 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007168.1 NP_001007169.1 3 - 4 418 88
XM_005171273.4 XP_005171330.1 3 - 4 425 88
GenBank mRNA BC079501.1 - 3 - 4 418 88

View Full Product Details