Assay Design



Gene Transcripts

Gene Symbol
ttc1
Entrez Gene ID
492510
Gene Name
tetratricopeptide repeat domain 1
Gene Aliases
im:7143277, zgc:101838
Chromosome Location
Chr.21: 30007566 - 30019626 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007382.1 NP_001007383.1 2 - 3 493 111
GenBank mRNA BC085453.1 - 2 - 3 493 111
BC164119.1 - 1 - 2 442 111

View Full Product Details