Assay Design



Gene Transcripts

Gene Symbol
capn2a
Entrez Gene ID
541374
Gene Name
calpain 2, (m/II) large subunit a
Gene Aliases
zgc:113590
Chromosome Location
Chr.13: 42728570 - 42756566 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013501.2 NP_001013519.1 3 - 4 508 64
GenBank mRNA BC090435.1 - 3 - 4 512 64

View Full Product Details