Assay Design



Gene Transcripts

Gene Symbol
enpp6
Entrez Gene ID
791915
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 6
Gene Aliases
E-NPP 6, cb1028, sb:cb727, zgc:103605
Chromosome Location
Chr.14: 4048134 - 4076480 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013527.1 NP_001013545.1 7 - 8 1248 132
GenBank mRNA BC090903.1 - 7 - 8 1248 132
BC165573.1 - 7 - 8 1149 132

View Full Product Details