Assay Design



Gene Transcripts

Gene Symbol
ctsk
Entrez Gene ID
550475
Gene Name
cathepsin K
Gene Aliases
wu:fa95f03, wu:fb08b05, zgc:110367
Chromosome Location
Chr.16: 29492994 - 29502549 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017778.1 NP_001017778.1 7 - 8 980 60
GenBank mRNA BC092901.1 - 7 - 8 980 60
BC164579.1 - 6 - 7 936 60

View Full Product Details