Assay Design



Gene Transcripts

Gene Symbol
hmbsb
Entrez Gene ID
338290
Gene Name
hydroxymethylbilane synthase, b
Gene Aliases
PBGD, hmbsl, id:ibd5004, im:7140060, zgc:110690
Chromosome Location
Chr.15: 23511403 - 23523222 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024388.1 NP_001019559.1 12 - 13 948 61
XM_005157569.4 XP_005157626.1 12 - 13 931 61
GenBank mRNA BC095373.1 - 12 - 13 948 61

View Full Product Details