Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024415.2 | NP_001019586.1 | 1 - 2 | 135 | 69 |
GenBank mRNA | BC095631.1 | - | 1 - 2 | 137 | 69 |
BC154355.1 | - | 1 - 2 | 134 | 69 | |
BC164758.1 | - | 1 - 2 | 102 | 69 |