Assay Design



Gene Transcripts

Gene Symbol
cdc37l1
Entrez Gene ID
569357
Gene Name
cell division cycle 37-like 1
Gene Aliases
im:7139934, wu:fa92d07, wu:fk28f10, zgc:171475
Chromosome Location
Chr.10: 10972795 - 10997564 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109826.2 NP_001103296.2 4 - 5 700 134
GenBank mRNA BC152570.1 - 4 - 5 700 134

View Full Product Details