Assay Design



Gene Transcripts

Gene Symbol
kri1
Entrez Gene ID
402800
Gene Name
KRI1 homolog
Gene Aliases
flj12949l, kri1l
Chromosome Location
Chr.3: 19336501 - 19355200 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002041.1 NP_001002041.1 16 - 17 1741 78
GenBank mRNA AY648765.1 - 16 - 17 1741 78
BC163908.1 - 16 - 17 1730 78

View Full Product Details