Assay Design



Gene Transcripts

Gene Symbol
mtg1
Entrez Gene ID
100150980
Gene Name
mitochondrial ribosome-associated GTPase 1
Gene Aliases
gtpbpl, zgc:109944, zgc:86745
Chromosome Location
Chr.13: 21674445 - 21682873 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002140.1 NP_001002140.1 8 - 9 735 121
NM_001144056.1 NP_001137528.1 5 - 6 365 121
XM_009307279.3 XP_009305554.1 8 - 9 705 121
XM_021480487.1 XP_021336162.1 8 - 9 693 121
GenBank mRNA BC071417.1 - 8 - 9 735 121
BC171457.1 - 5 - 6 365 121

View Full Product Details