Assay Design



Gene Transcripts

Gene Symbol
dcun1d2b
Entrez Gene ID
415246
Gene Name
DCN1, defective in cullin neddylation 1, domain containing 2b
Gene Aliases
dcun1d2, fj80d04, wu:fj80d04, zgc:86658
Chromosome Location
Chr.1: 208177 - 214921 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002156.2 NP_001002156.2 6 - 7 723 95
GenBank mRNA BC071344.1 - 6 - 7 95

View Full Product Details