Assay Design



Gene Transcripts

Gene Symbol
pqbp1
Entrez Gene ID
796523
Gene Name
polyglutamine binding protein 1
Gene Aliases
pqbp1l, si:dz150f13.4, zgc:92646
Chromosome Location
Chr.8: 36582728 - 36589390 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002435.1 NP_001002435.1 4 - 5 359 69
GenBank mRNA BC076131.1 - 4 - 5 359 69

View Full Product Details