Assay Design



Gene Transcripts

Gene Symbol
ftr24
Entrez Gene ID
110437704
Gene Name
finTRIM family, member 24
Gene Aliases
zgc:114036
Chromosome Location
Chr.2: 37294 - 45812 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001365968.1 NP_001352897.1 2 - 3 720 86
GenBank mRNA BC098890.1 - 2 - 3 618 86

View Full Product Details