Assay Design



Gene Transcripts

Gene Symbol
drgx
Entrez Gene ID
560399
Gene Name
dorsal root ganglia homeobox
Gene Aliases
prrxl1
Chromosome Location
Chr.13: 30911684 - 30918309 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037105.1 NP_001032182.1 2 - 3 161 79
XM_005156850.4 XP_005156907.1 3 - 4 759 79
GenBank mRNA AY753211.1 - 2 - 3 161 79

View Full Product Details