Assay Design



Gene Transcripts

Gene Symbol
gatd1
Entrez Gene ID
100148360
Gene Name
glutamine amidotransferase like class 1 domain containing 1
Gene Aliases
pddc1, zgc:123196
Chromosome Location
Chr.25: 3801227 - 3808635 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037118.1 NP_001032195.1 4 - 5 542 62
GenBank mRNA BC085644.1 - 3 - 4 1922 62
BC107609.1 - 4 - 5 542 62

View Full Product Details